Cobalt(II,III) oxide ≥99.9985% (Co basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

27-0490-25G 27-0490-5G
100202-388EA 365.17 USD
100202-388 100200-444
Cobalt(II,III) oxide ≥99.9985% (Co basis), PURATREM
Cobalt (II,III) oxide
Formula: Co₃O₄
MW: 240.8 g/mol
Melting Pt: 895
Density: 6.005 g/cm³ (20 °C)
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
Merck Index: 13,02454

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR