Iron(II) sulfide ≥99.9% (Fe basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ferrous sulphide, Ferrous sulfide

26-2635-10G 26-2635-50G
100206-512EA 111.82 USD
100206-512 100207-248
Iron(II) sulfide ≥99.9% (Fe basis)
Iron (II) sulphide
Formula: FeS
Melting Pt: 1195 °C
Density: 4.694 g/cm³ (20 °C)
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9,III
Merck Index: 12,04106

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR