Manganese ≥99.9%, chips

Supplier: STREM CHEMICALS INC MS
Danger

25-0050-2000G 25-0050-500G
100197-448EA 260.43 USD
100197-448 100208-862
Manganese ≥99.9%, chips
Manganese
Formula: Mn
Boiling Pt: 1900 °C
Melting Pt: 1260 °C
Density: 7.20
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3178
ADR: 4.1,II
Merck Index: 12,05762

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR