Vanadium ≥99.5%, foil
Supplier: STREM CHEMICALS INC MS
23-0050-100X50MM
23-0050-100X200MM
100198-020EA
246.28
USD
100198-020
100198-018
Vanadium ≥99.5%, foil
Vanadium
Formula:
V Boiling Pt: 3000 °C Melting Pt: 1717 °C Density: 6.11 |
MDL Number:
MFCD00011453 CAS Number: 7440-62-2 EINECS: 231-171-1 Merck Index: 12,10053 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...