Potassium ≥99.95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Breakseal ampoule
19-1990-1G
19-1990-5G
100200-882EA
157.11
USD
100200-882
100210-618
Potassium ≥99.95%
Potassium
Formula:
K Boiling Pt: 760 °C Melting Pt: 63.7 °C Density: 0.86 Storage Temperature: Ambient |
MDL Number:
MFCD00133776 CAS Number: 7440-09-7 EINECS: 231-119-8 UN: 2257 ADR: 4.3,I Merck Index: 12,07763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...