Tris(4-fluorophenyl)phosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

15-7890-1G 15-7890-5G
100201-506EA 84.92 USD
100201-506 100210-616
Tris(4-fluorophenyl)phosphine ≥99%
Tris(4-fluorophenyl)phosphine

Phosphine

Formula: C₁₈H₁₂F₃P
MW: 316.26 g/mol
Melting Pt: 80…81 °C
MDL Number: MFCD00013553
CAS Number: 18437-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR