Tripropylphosphine ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tri-n-propylphosphine

15-7050-50G 15-7050-10G
100202-640EA 1412.55 USD
100202-640 100196-116
Tripropylphosphine ≥95%
Tripropylphosphine

Phosphine

Formula: C₉H₂₁P
Boiling Pt: 185…188 °C
Density: 0,807
Flash Pt: 144 °F
MDL Number: MFCD00015217
CAS Number: 2234-97-1
EINECS: 218-786-0
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR