Trioctylphosphine oxide ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide
15-6661-100G
15-6661-25G
100208-622EA
193.91
USD
100208-622
100196-110
Trioctylphosphine oxide ≥99%
Trioctylphosphine oxide
Nanomaterials, Surfactants for Nano Synthesis
Formula:
C₂₄H₅₁OP Boiling Pt: 409 °C (1013 hPa) Melting Pt: 51…52 °C Density: 0,88 Flash Pt: >230 °F |
MDL Number:
MFCD00002083 CAS Number: 78-50-2 EINECS: 201-121-3 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...