Tricyclohexylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-6150-5G 15-6150-25G
100197-422EA 99.08 USD
100197-422 100196-918
Tricyclohexylphosphine ≥97%
Tricyclohexylphosphine

Phosphine

Formula: C₁₈H₃₃P
Boiling Pt: 132…134 °C (0.02 mmHg)
Melting Pt: 76…78 °C
Density: 0,9 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003853
CAS Number: 2622-14-2
EINECS: 220-069-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR