Triethylsilane ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
14-7750-25G
14-7750-100G
100200-408EA
123.14
USD
100200-408
100203-732
Triethylsilane ≥99%
Triethylsilane
Formula:
C₆H₁₆Si Boiling Pt: 107…108 °C Melting Pt: –156.9 °C Density: 0,728 Flash Pt: 25 °F Storage Temperature: Refrigerator |
MDL Number:
MFCD00009018 CAS Number: 617-86-7 EINECS: 210-535-3 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...