Sodium bis(trimethylsilyl)amide ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: HMDS sodium salt, N-Sodiohexamethyldisilazane, Sodium bis(trimethylsilyl)amide, HMDS-Na

11-1500-25G 11-1500-5G
100198-576EA 236.37 USD
100198-576 100203-950
Sodium bis(trimethylsilyl)amide ≥95%
Sodium bis(trimethylsilyl)amide
Formula: C₆H₁₈NNaSi₂
Density: 1.065 g/cm³ (20 °C)
MDL Number: MFCD00009835
CAS Number: 1070-89-9
EINECS: 213-983-8
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR