Grip Tight™ Circuit Breaker Lockouts, Master Lock
Supplier: Safety & Industrial Supplies
470-491B 470-493B
300081-482EA 27.97 USD
300081-482 300052-976
Grip Tight™ Circuit Breaker Lockouts, Master Lock
Security Locks
Secure and simple mounting for a solid lockout.
  • Grips tight with simple thumb turn and clamping handle
  • Accept all American Lock and Master Lock safety padlock shackle and lockout hasp diameters
  • Durable construction with powder coated steel and reinforced polymer allows operation in harsh environments