DALLAS Protective Eyewear, Crews®, ORS Nasco
Supplier: Safety & Industrial Supplies
135-DL110AF 135-DL210AF 135-DL110 135-DL310AF
300006-926EA 11.73 USD
300006-926 300061-496 300006-925 300061-506
DALLAS Protective Eyewear, Crews®, ORS Nasco
Safety Glasses
These eyewear has unique nosebridge with gel nosepad.
  • Bayonet temples
  • Full length non-slip temple sleeves
  • Wrap around lens design
Duramass® scratch-resistant lens coating with polycarbonate lens material.
Certifications: Meets or Exceeds ANSI Z87.1+ Standard.