Drum Handling Trucks, Harper Trucks
Supplier: Safety & Industrial Supplies
338-76A59 338-76A59P 338-9268-63 338-7889 338-7559 338-7559P
300079-788EA 977.51 USD
300079-788 300079-790 300079-794 300000-236 300068-634 300079-786
Drum Handling Trucks, Harper Trucks
Carts
Design enables one person to quickly, easily and safely move drums weighing up to 1000 or 1200 lbs.
  • Lifting toes create a cradle for easy break-over
  • Handles drums from 21 to 43 inches high
  • Replaceable chime hook and lifting toes ensure a long product life
  • Spring-loaded swing axle keeps truck in upright position when not in use
  • Also available with WH87 and WH89P wheels