Torqueplus™ Metric 12-Point Offset Box Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
577-1053M 577-1057M 577-1058M 577-1065M 577-1051M 577-1050M 577-1061M 577-1064M 577-1069M 577-1072M 577-1073M 577-1077M 577-1079M
300023-344EA 32.59 USD
300023-344 300023-346 300023-348 300023-354 300023-342 300023-356 300023-340 300023-350 300023-352 300023-358 300023-362 300023-364 300023-366 300023-368
Torqueplus™ Metric 12-Point Offset Box Wrenches, Proto®, ORS Nasco
Wrenches
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Reduces risk of fastener rounding
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Applies more torque to fastener