Thermo Scientific™ MAS™ Liquimmune Immunoassay Control, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
Save time with ready-to-use, liquid assayed MAS™ Liquimmune™ Immunoassay Controls
Features three significant levels. Human serum matrix. Assayed values available for most automated instruments.
Analytes: 17-a-hydroxyprogesterone, Acetaminophen, AFP, Aldosterone, Amikacin, Benzodiazepines, Carbamazepine, CEA, CK-MB, Cortisol, DHEA-Sulfate, Digoxin, Disopyramide, Estradiol, Ethosuximide, Ferritin, Folate, FSH, Gentamicin, Growth Hormone, ?-hCG, hCG, Homocysteine, IgA, IgE, IgG, IgM, LD-1, LH, Lidocaine, Lithium, NAPA, Phenobarbital, Phenytoin, Primidone, Procainamide, Progesterone, Prolactin, PSA, PSA Free, Quinidine, Salicylate, T Uptake, T3, T3 Free, T4, Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, TSH, Valproic Acid, Vancomycin, Vitamin B12 (refer to package insert for specific analyte and stability claims).
These multi-analyte controls are designed to monitor immunoassay test procedures on most automated instruments and are available in a convenient Vista™ vial format.
Ordering information: Available in Vista™ vial format, Multi-analyte control to help monitor immunoassay methods.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)