Thermo Scientific™ MAS™ Alcohol and Ammonia Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
AAC-MP
10014443
89130-580EA
423.03
USD
89130-580
10154-172
Thermo Scientific™ MAS™ Alcohol and Ammonia Controls, Thermo Scientific
Controls and Calibration Verification Materials
Monitor alcohol and/or ammonia in human serum with bi-level, assayed liquid MAS™ Alcohol and Ammonia Controls
Human-serum based matrix; Normal ammonia values in level 1; Assayed values available for most major analyzers.
Analytes: Alcohol, Ammonia
Available in a convenient Vista™ vial format, this ready-to-use product is packed in convenient multipacks.
Ordering information: Available in Vista™ vial format, Multipack (3 each of Levels 1 and 2).
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)