FILTER SAMPLER® Blood Serum Dispense Filter System, Porex
Supplier: Porex
6436 6437
28295-542CS 2244.73 USD
28295-542 28295-544
FILTER SAMPLER® Blood Serum Dispense Filter System, Porex
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Three-in-one filtration system filters, pipets, and stores samples
Restricted opening provides effective specimen closures and helps protect personnel. The dispense filter maintains sample integrity and eliminates the use of transfer pipets, add-on products, and closures. Use the Dispense Filter System model for glass collection tubes. Use the Dispense+ Serum Filter model specifically for 13mm plastic collection tubes.