Celite® Hyflo Super-cel®, filter aid
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22658-36
B22658-0E
AAB22658-36EA
57.76
USD
AAB22658-36
AAB22658-0E
Celite® Hyflo Super-cel®, filter aid
Celite® Hyflo® Super Cel, filter aid
CAS No.: 61790-53-2
Odor: Odorless
Merck Reference: 14,4973
MDL No.: MFCD00132803
Boiling Pt:
2200 °C (1013 hPa) Melting Pt: 1710 °C Density: 2.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00132803 CAS Number: 68855-54-9 EINECS: 319-127-6 Merck Index: 14,4973 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...