Diphenylcyclopropenone, (max. 5% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one

B21954-06 B21954-03 B21954-14
AAB21954-06EA 196 USD
AAB21954-06 AAB21954-03 AAB21954-14
Diphenylcyclopropenone, (max. 5% H₂O) 99% (dry weight)
Diphenylcyclopropenone

Purity: 99% CAS No.: 886-38-4 Molecular Formula: C15H10O Formula Weight: 206.24 Melting Point: 119-121° Beilstein Reference: 608049 Merck Reference: 14,3308 MDL No.: MFCD00001311

Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 407
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00001311
CAS Number: 886-38-4
EINECS: 212-948-4
Merck Index: 14,3308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR