N-Methyl-2-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

A19538-06 A19538-14 A19538-22
AAA19538-06EA 63.9 USD
AAA19538-06 AAA19538-14 AAA19538-22
N-Methyl-2-nitroaniline 98%
N-Methyl-2-nitroaniline

Purity: 98% CAS No.: 612-28-2 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Flash Point: 110°(230°F) Odor: Characteristic UN No.: 2811 MDL No.: MFCD00007090

Formula: C₇H₈N₂O₂
Boiling Pt: 158 °C (18 -rr)
Melting Pt: 33…36 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007090
CAS Number: 612-28-2
EINECS: 210-303-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR