Isonicotinamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Pyridinecarboxylic acid amide
A18190-22
A18190-18
A18190-30
A18190-36
AAAA18190-22EA
64.21
USD
AAAA18190-22
AAA18190-18
AAA18190-30
AAAA18190-36
Isonicotinamide 99%
Isonicotinamide
Purity: 99%
CAS No.: 1453-82-3
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Melting Point: 155-158°
Beilstein Reference: 2173
Formula:
C₆H₆N₂O MW: 122.13 g/mol Boiling Pt: 334 Melting Pt: 155…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006432 CAS Number: 1453-82-3 EINECS: 215-926-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...