Cellulose, microcrystalline

Supplier: Thermo Scientific Chemicals

Synonyms: Microcrystallinecellulose

A17730-36 A17730-0C A17730-0E
AAA17730-36EA 95.78 USD
AAA17730-36 AAA17730-0C AAA17730-0E
Cellulose, microcrystalline
Cellulose

CAS No.: 9004-34-6 Physical Form: Powder Appearance: White Merck Reference: 14,1965 MDL No.: MFCD00081512

Formula: (C₁₂H₂₂O₁₁)n
Melting Pt: 260…270 °C
Density: 1.27 to 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9
Merck Index: 14,1965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR