Chlorotriethylsilane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Triethylchlorosilane
A15547-09
A15547-18
A15547-30
AAA15547-09EA
70
USD
AAA15547-09
AAA15547-18
AAA15547-30
Chlorotriethylsilane 98+%
Chlorotriethylsilane
Purity: 98+%
CAS No.: 994-30-9
Molecular Formula: C6H15ClSi
Formula Weight: 150.73
Flash Point: 29°(84°F)
Refractive Index: 1.4310
Odor: Pungent
Melting Point: ca -75°
Boiling Point: 145-147°
Beilstein Reference: 1732860
UN No.: 2986
MDL No.: MFCD00000507
Formula:
C₆H₁₅ClSi Boiling Pt: 145…147 °C Melting Pt: ∼ 75 °C Density: 0.899 Flash Pt: 29 °C (84 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000507 CAS Number: 994-30-9 EINECS: 213-615-6 UN: 2986 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...