α,α,α-Trifluoro-p-toluic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Trifluoromethyl)benzoic acid
A14494-14
A14494-06
AAA14494-22EA
886.92
USD
AAA14494-22
AAA14494-14
AAAA14494-06
α,α,α-Trifluoro-p-toluic acid 98+%
α,α,α-Trifluoro-p-toluic acid
Formula:
C₈H₅F₃O₂ MW: 190.12 g/mol Boiling Pt: 247 °C/753 mm Melting Pt: 220…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002562 CAS Number: 455-24-3 EINECS: 207-242-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...