Pentafluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13948-22
A13948-14
A13948-06
AAA13948-22EA
328
USD
AAA13948-22
AAA13948-14
AAA13948-06
Pentafluorobenzene 98+%
Pentafluorobenzene
Purity: 98+%
CAS No.: 363-72-4
Molecular Formula: C6HF5
Formula Weight: 168.06
Flash Point: 13°(55°F)
Refractive Index: 1.3910
Melting Point: -48°
Boiling Point: 84-85°
Beilstein Reference: 1911549
UN No.: 1993
MDL No.: MFCD00000286
Formula:
C₆HF₅ Boiling Pt: 84…85 °C Melting Pt: –48 °C Density: 1.518 Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000286 CAS Number: 363-72-4 EINECS: 206-658-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...