Bromoacetonitrile 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13933-18
A13933-30
A13933-09
AAA13933-18EA
132
USD
AAA13933-18
AAAA13933-30
AAA13933-09
Bromoacetonitrile 96%
Bromoacetonitrile
Purity: 96%
CAS No.: 590-17-0
Molecular Formula: BrCH2CN
Formula Weight: 119.95
Flash Point: 81°(178°F)
Refractive Index: 1.4800
Boiling Point: 150°
Beilstein Reference: 956569
UN No.: 3276
MDL No.: MFCD00001884
Formula:
BrCNCH₂ MW: 119.95 g/mol Boiling Pt: 150 Melting Pt: -36 Density: 1.722 Flash Pt: 81 °C (178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001884 CAS Number: 590-17-0 EINECS: 209-672-1 UN: 2922 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...