Flavone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Phenylchromen-4-one

A13627-06 A13627-14 A13627-03
AAA13627-06EA 84.3 USD
AAA13627-06 AAA13627-14 AAA13627-03
Flavone 99%
Flavone

Purity: 99% CAS No.: 525-82-6 Molecular Formula: C15H10O2 Formula Weight: 222.24 Odor: Odorless Melting Point: 95-98° Beilstein Reference: 157598 Merck Reference: 14,4092 MDL No.: MFCD00006825

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 180…185 °C (1 mmHg)
Melting Pt: 96
Storage Temperature: Ambient
MDL Number: MFCD00006825
CAS Number: 525-82-6
EINECS: 208-383-8
Merck Index: 14,4092

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR