4-Bromobiphenyl (PBB No. 3) 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl
A12638-30
A12638-09
A12638-18
AAA12638-30EA
464
USD
AAA12638-30
AAA12638-09
AAA12638-18
4-Bromobiphenyl (PBB No. 3) 98+%
4-Bromobiphenyl (PBB No. 3)
Purity: 98+%
CAS No.: 92-66-0
Molecular Formula: C12H9Br
Formula Weight: 233.12
Flash Point: 143°(289°F)
Melting Point: 88-92°
Boiling Point: 310°
Beilstein Reference: 1907453
MDL No.: MFCD00000100
Formula:
C₁₂H₉Br MW: 233.11 g/mol Boiling Pt: 310 °C Melting Pt: 88…92 °C Density: 1.615 g/cm³ (20 °C) Flash Pt: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000100 CAS Number: 92-66-0 EINECS: 202-176-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...