Cyclopropylméthanol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclopropyl carbinol, Cyclopropanemethanol

A12567-14 A12567-22 A12567-06
AAA12567-14EA 68.8 USD
AAA12567-14 AAA12567-22 AAA12567-06
Cyclopropylméthanol 98%
Cyclopropylméthanol

Purity: 98% CAS No.: 2516-33-8 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: 35°(95°F) Refractive Index: 1.4330 Odor: Mild Melting Point: -60° Boiling Point: 123-124° Beilstein Reference: 1846846 UN No.: 1987 MDL No.: MFCD00001309

Formula: C₄H₈O
Boiling Pt: 123…124 °C
Melting Pt: < –60 °C
Density: 0.904
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001309
CAS Number: 2516-33-8
EINECS: 219-735-5
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR