2-Furonitrile 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12214-06 A12214-14
AAA12214-06EA 88.4 USD
AAA12214-06 AAA12214-14
2-Furonitrile 98+%
2-Furonitrile

MDL: MFCD00003223 Beilstein Registry No.: 107033


Purity: 98+% CAS No.: 617-90-3 Molecular Formula: C5H3NO Formula Weight: 93.09 Flash Point: 35°(95°F) Refractive Index: 1.4790 Odor: Recognizable Boiling Point: 146-148° Beilstein Reference: 107033 UN No.: 1992 MDL No.: MFCD00003223

Formula: C₅H₃NO
Boiling Pt: 146…148 °C
Density: 1.062
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003223
CAS Number: 617-90-3
EINECS: 210-537-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR