Theobromine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dimethylxanthine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3,7-dimethylpurine

A11861-18 A11861-30 A11861-22
AAA11861-18EA 141.3 USD
AAA11861-18 AAA11861-30 AAAA11861-22
Theobromine 99%
Theobromine

Phosphosdiesterase inhibitor


Purity: 99% CAS No.: 83-67-0 Molecular Formula: C7H8N4O2 Formula Weight: 180.17 Melting Point: 345-350° Beilstein Reference: 16464 Merck Reference: 14,9282 MDL No.: MFCD00022830


Phosphosdiesterase inhibitor

Formula: C₇H₈N₄O₂
MW: 180.17 g/mol
Melting Pt: 345…350 °C
Density: 1.50
Storage Temperature: Ambient
MDL Number: MFCD00022830
CAS Number: 83-67-0
EINECS: 201-494-2
Merck Index: 14,9282

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR