Hexafluorobenzene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Perfluorobenzene

A11500-09 A11500-18 A11500-30
AAA11500-09EA 57.13 USD
AAA11500-09 AAA11500-18 AAA11500-30
Hexafluorobenzene 99%
Hexafluorobenzene

MDL: MFCD00000288 Beilstein Registry No.: 1683438


Purity: 99% CAS No.: 392-56-3 Molecular Formula: C6F6 Formula Weight: 186.06 Flash Point: 10°(50°F) Refractive Index: 1.3780 Odor: Aromatic Melting Point: 4-5° Boiling Point: 80-81° Beilstein Reference: 1683438 Merck Reference: 14,4686 UN No.: 1993 MDL No.: MFCD00000288

Formula: C₆F₆
MW: 186.06 g/mol
Boiling Pt: 80…81 °C
Melting Pt: 4…5 °C
Density: 1.616
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000288
CAS Number: 392-56-3
EINECS: 206-876-2
UN: 1993
ADR: 3,III
Merck Index: 14,4686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR