Caprylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Octanoic acid, n-Octanoic Acid, Octoic acid
A11149-AP
A11149-AC
A11149-0F
A11149-0D
AAA11149-APEA
58.86
USD
AAA11149-AP
AAAA11149-AC
AAA11149-0F
AAA11149-0D
Caprylic acid 98+%
Caprylic acid
MDL: MFCD00004429
Beilstein Registry No.: 1747180
Purity: 98+%
CAS No.: 124-07-2
Molecular Formula: C8H16O2
Formula Weight: 144.21
Flash Point: 132°(269°F)
Physical Form: Liquid
Density: 0.910
Refractive Index: 1.4275
Appearance: Colorless
Odor: Rancid
pH: 4
Melting Point: 16-17°
Boiling Point: 236-238°
Beilstein Reference: 1747180
Merck Reference: 14,1765
UN No.: 3265
MDL No.: MFCD00004429
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Boiling Pt: 236…238 °C Melting Pt: 16…17 °C Density: 0.910 Flash Pt: 132 °C (269 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004429 CAS Number: 124-07-2 EINECS: 204-677-5 UN: 3265 ADR: 8,III Merck Index: 14,1765 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...