Diphenyltin dichloride ≥95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dichlorodiphenylstannane

071139-04 071139-09 071139-18
AA71139-04EA 19.1 USD
AA71139-04 AA71139-09 AA71139-18
Diphenyltin dichloride ≥95%
Diphenyltin dichloride

MDL: MFCD00000516


Purity: 95% CAS No.: 1135-99-5 Molecular Formula: C12H10Cl2Sn Formula Weight: 343.81 Flash Point: 150°(302°F) Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White Melting Point: 41-43° Boiling Point: 333-337° dec. MDL No.: MFCD00000516

Formula: C₁₂H₁₀Cl₂Sn
Boiling Pt: 333…337 °C (dec.)
Melting Pt: 41…43 °C
Flash Pt: >150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000516
CAS Number: 1135-99-5
EINECS: 214-496-3
UN: 3146
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR