Fullerene C70 99+%, powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Buckminster-Fullerene C70, [5,6]-Fullerene-C70
042600-03
042600-MD
AA42600-03EA
1034
USD
AA42600-03
AA42600-MD
Fullerene C70 99+%, powder
Fullerene C70
MDL: MFCD00282904
Purity: 99+%
CAS No.: 115383-22-7
Physical Form: Powder
Density: 1.7
Appearance: Grey/black
Boiling Point: 500-600° subl.
UN No.: 1325
MDL No.: MFCD00282904
Formula:
C₇₀ MW: 840.77 g/mol Boiling Pt: 500…600 °C (subl.) Melting Pt: > 280 °C Density: 1.7 Storage Temperature: Ambient |
MDL Number:
MFCD00282904 CAS Number: 115383-22-7 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...