n-Tetracosane 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-tetracosane

042462-14 042462-22 042462-06
AA42462-14EA 47.3 USD
AA42462-14 AA42462-22 AA42462-06
n-Tetracosane 99%
n-Tetracosane

MDL: MFCD00009352


Purity: 99% CAS No.: 646-31-1 Molecular Formula: CH3(CH2)22CH3 Formula Weight: 338.66 Flash Point: 110°(230°F) Physical Form: Crystalline Melting Point: 49-52° Boiling Point: 391° MDL No.: MFCD00009352

Formula: H₃C(CH₂)₂₂CH₃
MW: 338.66 g/mol
Boiling Pt: 391.3
Melting Pt: 49…52 °C
Density: 0.7787 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009352
CAS Number: 646-31-1
EINECS: 211-474-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR