n-Octacosane 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-octacosane

042460-30 042460-09 042460-18
AA42460-30EA 342 USD
AA42460-30 AA42460-09 AA42460-18
n-Octacosane 99%
n-Octacosane

MDL: MFCD00009355


Purity: 99% CAS No.: 630-02-4 Molecular Formula: CH3(CH2)26CH3 Formula Weight: 394.77 Flash Point: 227°(441°F) Physical Form: Crystalline Melting Point: 61-63° Boiling Point: 440° MDL No.: MFCD00009355

Formula: H₃C(CH₂)₂₆CH₃
MW: 394.77 g/mol
Boiling Pt: 432
Melting Pt: 61…63 °C
Density: 0.8067 g/cm³ (20 °C)
Flash Pt: 227 °C (441 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009355
CAS Number: 630-02-4
EINECS: 211-125-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR