Acetone-[D₆] (100% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyl ketone-D6
42262-XF
42262-XF
042262-AA
42262-AA
AA42262-AAEA
162.31
USD
AA42262-AA
CAAA42262-XF
AA42262-XF
CAAA42262-AA
Acetone-[D₆] (100% D) (Isotopic) for NMR spectroscopy
Acetone-[D6]
Formula:
D₃CC(O)CD₃ MW: 64.03 g/mol Boiling Pt: 56 °C Melting Pt: –94 °C Density: 0.872 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044635 CAS Number: 666-52-4 EINECS: 211-563-9 UN: 1090 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...