Beryllium ≥99.8% (metals basis), foil, Thickness 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

041643-FI 041643-FF
AA41643-FIEA 2745 USD
AA41643-FI AA41643-FF
Beryllium ≥99.8% (metals basis), foil, Thickness 0.1 mm (0.004 in)
Beryllium

MDL: MFCD00134032


Purity: 99.8% CAS No.: 7440-41-7 Physical Form: ≈0.12g/25x25mm, Vacuum tight Odor: Odorless MDL No.: MFCD00134032

Formula: Be
Boiling Pt: 2970 °C (1013 hPa)
Melting Pt: 1278 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134032
CAS Number: 7440-41-7
EINECS: 231-150-7
Merck Index: 12,01201

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR