Polyvinylpyrrolidone, M.W. ∼ 8.000 (K16-18)

Supplier: Thermo Scientific Chemicals

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

041626-36 041626-22
AA41626-36EA 298 USD
AA41626-36 AA41626-22
Polyvinylpyrrolidone, M.W. ∼ 8.000 (K16-18)
Polyvinylpyrrolidone

A cryoprotectant, also useful in plant cell culture research


CAS No.: 9003-39-8 Molecular Formula: (C6H9NO)n Physical Form: Powder Odor: Odorless Melting Point: 130° Merck Reference: 14,7697 MDL No.: MFCD00149016


A cryoprotectant, also useful in plant cell culture research

Formula: (C₆H₉NO)n
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR