Iron ≥98%, spherical powder 1-3 µm

Supplier: Thermo Scientific Chemicals
Danger

040337-36 040337-95 040337-14 040337-22
AA40337-36EA 133 USD
AA40337-36 AA40337-95 AA40337-14 AA40337-22
Iron ≥98%, spherical powder 1-3 µm
Iron

MDL: MFCD00010999


Purity: 98+% CAS No.: 7439-89-6 Physical Form: Carbonyl iron powder Appearance: Gray to brown UN No.: 3089 MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1 II
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR