Lanthanum ≥99.9% (REO, rare earth oxide basis), rods, Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals
Danger

040292-DH 040292-DJ 040292-DM 040292-HF
AA40292-DHEA 234 USD
AA40292-DH AA40292-DJ AA40292-DM AA40292-HF
Lanthanum ≥99.9% (REO, rare earth oxide basis), rods, Diameter 6.35 mm (0.25 in)
Lanthanum

MDL: MFCD00011066


Purity: 99.9% CAS No.: 7439-91-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.20g/mm, Cast rod, diameter is approximate Appearance: Gray to silver UN No.: 3208 MDL No.: MFCD00011066

Formula: La
Boiling Pt: 3464 °C (1013 hPa)
Melting Pt: 920 °C
Density: ∼6,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR