Ammonium chloride ≥99.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium muriate, Salmiac

040193-A1 040193-A7
AA40193-A1EA 131.8 USD
AA40193-A1 AA40193-A7
Ammonium chloride ≥99.5% ACS
Ammonium chloride

MDL: MFCD00011420


Purity: 99.5% min CAS No.: 12125-02-9 Molecular Formula: NH4Cl Formula Weight: 53.49 Physical Form: Crystalline Density: ≈1.53 Refractive Index: 1.642 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 340° subl. Merck Reference: 14,509 MDL No.: MFCD00011420

Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335 (subl.)
Density: 1.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 14,509
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.005%
Residue after ignition 0.01%
PO11 2 ppm
SO3 0.002%
Ca 0.001%
MG 5 ppm
Heavy Metals (as Pb) 5 ppm max.
Fe 2 ppm
pH of a 5% solution 4.5-5.5 at 25°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR