Boron 94-96%, amorphous powder APS <5 µm (max. 3% Mg)
Supplier: Thermo Scientific Chemicals
|
Danger
|
039680-30
039680-18
AA39680-30EA
496
USD
AA39680-30
AA39680-18
Boron 94-96%, amorphous powder APS <5 µm (max. 3% Mg)
Boron
MDL: MFCD00134034
Purity: 94-96%
CAS No.: 7440-42-8
Physical Form: Oxygen is the major impurity
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00134034
Formula:
B Density: 1.73 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134034 CAS Number: 7440-42-8 EINECS: 231-151-2 UN: 3178 ADR: 4.1,II Merck Index: 13,01333 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...