Carbon disulfide ≥99.8% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

038993-K2 038993-M1
AA38993-K2EA 220.47 USD
AA38993-K2 AA38993-M1
Carbon disulfide ≥99.8% for HPLC
Carbon disulphide

MDL: MFCD00011321 Notes: Suitable for spectrophotometry


Purity: 99.8+% CAS No.: 75-15-0 Molecular Formula: CS2 Formula Weight: 76.14 Flash Point: -30°(-22°F) Physical Form: Liquid Density: 1.2632 Refractive Index: 1.6270 Odor: Unpleasant Melting Point: -112° Boiling Point: 46° Merck Reference: 14,1811 UN No.: 1131 MDL No.: MFCD00011321

Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46 °C
Melting Pt: –112 °C
Density: 1.2632
Flash Pt: –30 °C (–22 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 14,1811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR