Bismuth ≥99.999% (metals basis), spherical shots 1-5 mm (0.04-0.20 in)

Supplier: Thermo Scientific Chemicals

038619-18 038619-09
AA38619-18EA 653 USD
AA38619-18 AA38619-09
Bismuth ≥99.999% (metals basis), spherical shots 1-5 mm (0.04-0.20 in)
Bismuth

Purity: 99.999% CAS No.: 7440-69-9 Physical Form: Ampouled under argon Odor: Odorless MDL No.: MFCD00134033

Formula: Bi
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR