Glassy carbon, rods, Diameter 2 mm (0.08 in) Type 1

Supplier: Thermo Scientific Chemicals

038010-DJ 038010-DM 038010-DR
AA38010-DJEA 278 USD
AA38010-DJ AA38010-DM AA38010-DR
Glassy carbon, rods, Diameter 2 mm (0.08 in) Type 1
Glassy carbon

MDL: MFCD00133992


Physical Form: ≈0.05g/cm MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR