Glassy carbon, rods, Diameter 2 mm (0.08 in) Type 2

Supplier: Thermo Scientific Chemicals

037997-DJ 037997-DM 037997-DR
AA37997-DJEA 210.12 USD
AA37997-DJ AA37997-DM AA37997-DR
Glassy carbon, rods, Diameter 2 mm (0.08 in) Type 2
Glassy carbon

MDL: MFCD00133992


Physical Form: ≈0.04g/cm MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR