1,3-Dichloropropane ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylendichloride
036685-14
036685-22
AA36685-14EA
91.1
USD
AA36685-14
AA36685-22
1,3-Dichloropropane ≥95%
1,3-Dichloropropane
MDL: MFCD00000999
Purity: 95%
CAS No.: 142-28-9
Molecular Formula: Cl(CH2)3Cl
Formula Weight: 112.99
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 1.190
Refractive Index: 1.4481
Odor: Sweet
Boiling Point: 120°
UN No.: 1993
MDL No.: MFCD00000999
Formula:
ClH₂CCH₂CH₂Cl MW: 112.99 Boiling Pt: 120.4 Melting Pt: -99.5 Density: 1.190 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000999 CAS Number: 142-28-9 EINECS: 205-531-3 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...